Learn More
ABclonal Technology KHSRP Rabbit mAb
SDP
Catalog No. 5000135641
Encompass_Preferred
The KHSRP gene encodes a multifunctional RNA-binding protein implicated in a variety of cellular processes, including transcription, alternative pre-mRNA splicing, and mRNA localization (Min et al., 1997 [PubMed 9136930], Gherzi et al., 2004 [PubMed 15175153]).
Catalog No. 50-001-35641 Supplier ABclonal Technology Supplier No. A9075500UL
The KHSRP gene encodes a multifunctional RNA-binding protein implicated in a variety of cellular processes, including transcription, alternative pre-mRNA splicing, and mRNA localization (Min et al., 1997 [PubMed 9136930], Gherzi et al., 2004 [PubMed 15175153]).