Learn More
ABclonal Technology NCK1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A91291000UL
Encompass_Preferred
The protein encoded by this gene is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.
Catalog No. 50-001-35675