Learn More
ABclonal Technology Hemopexin (HPX) Rabbit mAb
SDP
Supplier:  ABclonal Technology A9133500UL
Encompass_Preferred
This gene encodes a plasma glycoprotein that binds heme with high affinity. The encoded protein is an acute phase protein that transports heme from the plasma to the liver and may be involved in protecting cells from oxidative stress.
Catalog No. 50-001-36940