Learn More
Selleck Chemical LLC A922500 5mg 959122-11-3
SDP
Supplier:  Selleck Chemical LLC S26745MG
Encompass_Preferred
A922500 is an inhibitor for human and mouse DGAT-1 with IC50 of 7 nM and 24 nM, respectively, good selectivity over related acyltransferases, hERG, and a panel of anti-targets. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2734