Learn More
Apexbio Technology LLC A922500, 5mg. Cas: 959122-11-3 MFCD: MFCD12828780. DGAT-1 inhibitor
SDP
Catalog No. 501904936
Encompass_Preferred
A922500 is a potent, active, selective and orally bioavailable inhibitor of diacylglycerol acyltransferase-1 (DGAT-1). It is a small molecular with the formula of C26H24N2O4 and molecular weight of 428.48. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4936 Supplier Apexbio Technology LLC Supplier No. A43825
A922500 is a potent, active, selective and orally bioavailable inhibitor of diacylglycerol acyltransferase-1 (DGAT-1). It is a small molecular with the formula of C26H24N2O4 and molecular weight of 428.48. Other sizes are available. Please inqury us for quote.