Learn More
ABclonal Technology USP22 Rabbit mAb
SDP
Catalog No. 5000135454
Encompass_Preferred
Enables enzyme binding activity, nuclear receptor coactivator activity, and thiol-dependent deubiquitinase. Contributes to H4 histone acetyltransferase activity. Involved in positive regulation of mitotic cell cycle, positive regulation of transcription, DNA-templated, and protein modification by small protein conjugation or removal. Part of SAGA complex.
Catalog No. 50-001-35454 Supplier ABclonal Technology Supplier No. A92611000UL
Enables enzyme binding activity, nuclear receptor coactivator activity, and thiol-dependent deubiquitinase. Contributes to H4 histone acetyltransferase activity. Involved in positive regulation of mitotic cell cycle, positive regulation of transcription, DNA-templated, and protein modification by small protein conjugation or removal. Part of SAGA complex.