Learn More
ABclonal Technology AP2S1 Rabbit mAb
SDP
Catalog No. 5000136140
Encompass_Preferred
One of two major clathrin-associated adaptor complexes, AP-2, is a heterotetramer which is associated with the plasma membrane. This complex is composed of two large chains, a medium chain, and a small chain. This gene encodes the small chain of this complex. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-36140 Supplier ABclonal Technology Supplier No. A92631000UL
One of two major clathrin-associated adaptor complexes, AP-2, is a heterotetramer which is associated with the plasma membrane. This complex is composed of two large chains, a medium chain, and a small chain. This gene encodes the small chain of this complex. Alternative splicing results in multiple transcript variants.