Learn More
ABclonal Technology MC5 Receptor Rabbit mAb
SDP
Catalog No. 5000134783
Encompass_Preferred
This gene encodes a member of the seven-pass transmembrane G protein-coupled melanocortin receptor protein family that stimulate cAMP signal transduction. The encoded protein is a receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone and is suggested to play a role in sebum generation.
Catalog No. 50-001-34783 Supplier ABclonal Technology Supplier No. A93311000UL
This gene encodes a member of the seven-pass transmembrane G protein-coupled melanocortin receptor protein family that stimulate cAMP signal transduction. The encoded protein is a receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone and is suggested to play a role in sebum generation.