Learn More
ABclonal Technology PPP6C Rabbit mAb
SDP
Catalog No. 5000134784
Encompass_Preferred
This gene encodes the catalytic subunit of protein phosphatase, a component of a signaling pathway regulating cell cycle progression. Splice variants encoding different protein isoforms exist. The pseudogene of this gene is located on chromosome X.
Catalog No. 50-001-34784 Supplier ABclonal Technology Supplier No. A9336500UL
This gene encodes the catalytic subunit of protein phosphatase, a component of a signaling pathway regulating cell cycle progression. Splice variants encoding different protein isoforms exist. The pseudogene of this gene is located on chromosome X.