Learn More
ABclonal Technology SAMM50 Rabbit mAb
SDP
Catalog No. 5000136411
Encompass_Preferred
This gene encodes a component of the Sorting and Assembly Machinery (SAM) of the mitochondrial outer membrane. The Sam complex functions in the assembly of beta-barrel proteins into the outer mitochondrial membrane.
Catalog No. 50-001-36411 Supplier ABclonal Technology Supplier No. A9355500UL
This gene encodes a component of the Sorting and Assembly Machinery (SAM) of the mitochondrial outer membrane. The Sam complex functions in the assembly of beta-barrel proteins into the outer mitochondrial membrane.