Learn More
Medchemexpress LLC HY-50709 10mg Medchemexpress, A939572 CAS:1032229-33-6 Purity:>98%
SDP
Catalog No. 502029331
Encompass_Preferred
Medchemexpress, HY-50709 10mg A939572 CAS:1032229-33-6 A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of <4 nM and 37 nM for mSCD1 and hSCD1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9331 Supplier Medchemexpress LLC Supplier No. HY5070910MG
Medchemexpress, HY-50709 10mg A939572 CAS:1032229-33-6 A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of <4 nM and 37 nM for mSCD1 and hSCD1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.