Learn More
Apexbio Technology LLC A939572 1032229-33-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063913
Encompass_Preferred
A939572 (CAS 1032229-33-6) is an orally bioavailable small-molecule inhibitor targeting stearoyl-CoA desaturase 1 (SCD1) an enzyme that catalyzes the formation of monounsaturated fatty acids involved in lipid metabolism and body weight regulation A939572 selectively inhibits human and mouse SCD1 with IC50 values of 37 nM and 4 nM respectively exhibiting no inhibitory effect on associated cofactors cytochrome b5 or cytochrome b5 reductase By specifically interacting with SCD1 A939572 serves as a pharmacological tool in metabolic research particularly in studies focused on obesity and diabetes
Catalog No. 50-000-63913 Supplier Apexbio Technology LLC Supplier No. B36075.1
 Restricted Item
Add to Cart
Add to Cart
A939572 (CAS 1032229-33-6) is an orally bioavailable small-molecule inhibitor targeting stearoyl-CoA desaturase 1 (SCD1) an enzyme that catalyzes the formation of monounsaturated fatty acids involved in lipid metabolism and body weight regulation A939572 selectively inhibits human and mouse SCD1 with IC50 values of 37 nM and 4 nM respectively exhibiting no inhibitory effect on associated cofactors cytochrome b5 or cytochrome b5 reductase By specifically interacting with SCD1 A939572 serves as a pharmacological tool in metabolic research particularly in studies focused on obesity and diabetes