Learn More
Medchemexpress LLC HY-50709 50mg , A939572 stearoyl-CoA desaturase (SCD) inhibitor;SCD-inhibitor CAS:1032229-33-6 Purity:98%
SDP
Catalog No. 501493691
Encompass_Preferred
Medchemexpress, HY-50709 50mg A939572 stearoyl-CoA desaturase (SCD) inhibitor;SCD-inhibitor CAS:1032229-33-6 Formula:C20H22ClN3O3 IC50: <4 nM (mSCD1), 37 nM (hSCD1) Purity:98% A939572 is a potent, and orally bioavailable SCD1 inhibitor with IC 50 values of <4 nM and 37 nM for mSCD1 and hSCD1 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3691 Supplier Medchemexpress LLC Supplier No. HY5070950MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-50709 50mg A939572 stearoyl-CoA desaturase (SCD) inhibitor;SCD-inhibitor CAS:1032229-33-6 Formula:C20H22ClN3O3 IC50: <4 nM (mSCD1), 37 nM (hSCD1) Purity:98% A939572 is a potent, and orally bioavailable SCD1 inhibitor with IC 50 values of <4 nM and 37 nM for mSCD1 and hSCD1 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.