Learn More
Selleck Chemical LLC A939572-5mg
SDP
Catalog No. 502389759
Encompass_Preferred
A939572 is a potent and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 of <4 nM and 37 nM for mSCD1 and hSCD1, respectively.
Catalog No. 50-238-9759 Supplier Selleck Chemical LLC Supplier No. S96075MG
A939572 is a potent and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 of <4 nM and 37 nM for mSCD1 and hSCD1, respectively.