Learn More
ABclonal Technology ENPP5 Rabbit mAb
SDP
Catalog No. 5000134832
Encompass_Preferred
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.
Catalog No. 50-001-34832 Supplier ABclonal Technology Supplier No. A9415500UL
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.