Learn More
Cayman Chemical A-943931 (hydrochloride hydrate), Purity: ≥98%, 25 mg
SDP
Catalog No. 502946977
Encompass_Preferred
A histamine H4 receptor antagonist (Ki = 3.8 nM); inhibits clobenpropit-induced scratching in mice (ED50 = 26 µmoles/kg, i.p.)
Catalog No. 50-294-6977 Supplier Cayman Chemical Supplier No. 3455425 MG
A histamine H4 receptor antagonist (Ki = 3.8 nM); inhibits clobenpropit-induced scratching in mice (ED50 = 26 µmoles/kg, i.p.)