Learn More
ABclonal Technology UQCRFS1 Rabbit mAb
SDP
Catalog No. 5000136220
Encompass_Preferred
Predicted to enable oxidoreductase activity. Involved in mitochondrial respiratory chain complex III assembly and respiratory electron transport chain. Located in mitochondrion. Part of mitochondrial respiratory chain complex III and mitochondrial respiratory chain complex IV. Implicated in mitochondrial complex III deficiency.
Catalog No. 50-001-36220 Supplier ABclonal Technology Supplier No. A95171000UL
Predicted to enable oxidoreductase activity. Involved in mitochondrial respiratory chain complex III assembly and respiratory electron transport chain. Located in mitochondrion. Part of mitochondrial respiratory chain complex III and mitochondrial respiratory chain complex IV. Implicated in mitochondrial complex III deficiency.