Learn More
ABclonal Technology TorsinA/TOR1A Rabbit mAb
SDP
Catalog No. 5000134884
Encompass_Preferred
The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.
Catalog No. 50-001-34884 Supplier ABclonal Technology Supplier No. A95791000UL
The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.