Learn More
ABclonal Technology GABARAPL2 Rabbit mAb
SDP
Catalog No. 5000135554
Encompass_Preferred
Enables ubiquitin protein ligase binding activity. Involved in negative regulation of proteasomal protein catabolic process and protein localization to endoplasmic reticulum. Located in Golgi membrane and autophagosome membrane.
Catalog No. 50-001-35554 Supplier ABclonal Technology Supplier No. A95851000UL
Enables ubiquitin protein ligase binding activity. Involved in negative regulation of proteasomal protein catabolic process and protein localization to endoplasmic reticulum. Located in Golgi membrane and autophagosome membrane.