Learn More
ABclonal Technology TOMM22 Rabbit mAb
SDP
Catalog No. 5000135333
Encompass_Preferred
The protein encoded by this gene is an integral membrane protein of the mitochondrial outer membrane. The encoded protein interacts with TOMM20 and TOMM40, and forms a complex with several other proteins to import cytosolic preproteins into the mitochondrion.
Catalog No. 50-001-35333 Supplier ABclonal Technology Supplier No. A96661000UL
The protein encoded by this gene is an integral membrane protein of the mitochondrial outer membrane. The encoded protein interacts with TOMM20 and TOMM40, and forms a complex with several other proteins to import cytosolic preproteins into the mitochondrion.