Learn More
Inorganic Ventures 1000µg/mL IRON 125mL
SDP
Catalog No. 50723065
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: Fe metal, Analyte: Fe 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-065 Supplier Inorganic Ventures Supplier No. AAFE1125ML
Add to Cart
Add to Cart
Matrix: Nitric Acid (HNO3), Starting Material: Fe metal, Analyte: Fe 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search