Learn More
Inorganic Ventures 1000µg/mL MANGANESE 500mL
SDP
Catalog No. 50723094
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Manganese for AA is a 500mL reference material set in a nitric acid matrix for stability. This manganese standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-094 Supplier Inorganic Ventures Supplier No. AAMN1500ML
Add to Cart
Add to Cart
Inorganic Ventures' 1000 ppm (µg/mL) Manganese for AA is a 500mL reference material set in a nitric acid matrix for stability. This manganese standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search