Learn More
Inorganic Ventures 1000µg/mL MOLYBDENUM 500mL
SDP
Catalog No. 50723096
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Molybdenum for AA is a 125mL certified reference material set in an ammonium hydroxide matrix for stability. This molybdenum standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-096 Supplier Inorganic Ventures Supplier No. AAMO1500ML
Add to Cart
Add to Cart
Inorganic Ventures' 1000 ppm (µg/mL) Molybdenum for AA is a 125mL certified reference material set in an ammonium hydroxide matrix for stability. This molybdenum standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search