Learn More
Inorganic Ventures 1000µg/mL SODIUM 125mL
SDP
Catalog No. 50723097
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Molybdenum for AA is a 500mL certified reference material set in an ammonium hydroxide matrix for stability. This molybdenum standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-097 Supplier Inorganic Ventures Supplier No. AANA1125ML
Add to Cart
Add to Cart
Inorganic Ventures' 1000 ppm (µg/mL) Molybdenum for AA is a 500mL certified reference material set in an ammonium hydroxide matrix for stability. This molybdenum standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search