Learn More
Inorganic Ventures 1000µg/mL URANIUM 500mL
SDP
Supplier:  Inorganic Ventures AAU1500ML
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Uranium for AA is a 500mL reference material set in a nitric acid matrix for stability. This uranium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-154