Learn More
Perkin Elmer US LLC AA and ICP Vent and Blower Kit - 230V
Catalog No. 502112795
Encompass_Preferred
Included in the kit are a rotary blower with capacitor and hardware, a vent to be located above the instrument, and an adapter to permit connection of the blower and vent with suitable metal tubing. The adapter and vent are made of stainless steel sheets. * Notice: Local electrical codes do not permit PerkinElmer Service Engineers to install the blower and vent assembly.
Catalog No. 50-211-2795 Supplier Perkin Elmer US LLC Supplier No. 03030448
Included in the kit are a rotary blower with capacitor and hardware, a vent to be located above the instrument, and an adapter to permit connection of the blower and vent with suitable metal tubing. The adapter and vent are made of stainless steel sheets. * Notice: Local electrical codes do not permit PerkinElmer Service Engineers to install the blower and vent assembly.