Learn More
Mopec AA Collection Swabs and Nitric Acid Dispenser Kit
SDP
Catalog No. 22446317 Shop All Mopec Products
Change view
Click to view available options
Quantity:
25/Cs.
Catalog No. Quantity
22-446-317 25/Cs.
1 options
Catalog No. 22-446-317 Supplier Mopec Supplier No. FS478
Add to Cart
Add to Cart
Designed for collection and screening of residue from a subject, the supplied treated swabs support sample recovery while a controlled nitric acid dispensing system supports consistent preparation. Together, the components provide materials needed to prepare collected residue for analysis.
Construction
  • (2) swabs supplied in each of the (5) polypropylene snap cap vials
  • Write-on labels
  • (1) 3 mL dispenser containing a 5% nitric acid solution
Performance
  • For collection of additional GSR from a site other than a suspect’s hand
Quantity
  • 25 per case
Specifications
Quantity 25/Cs.