Learn More
Beckman Coulter Calibration Standards Kit
Catalog No. NC1505340
Encompass
Calibration Standards Kit, includes: calibration blank and 0.250, 0.500 and 0.750 mg C/L sucrose calibration standards in RFID bottles.
Catalog No. NC1505340 Supplier Beckman Coulter Supplier No. FG7019202
Calibration Standards Kit, includes: calibration blank and 0.250, 0.500 and 0.750 mg C/L sucrose calibration standards in RFID bottles.