Learn More
Perkin Elmer US LLC AA Test Mix
Catalog No. 502113469
Encompass_Preferred
Recommended for use in checking sensitivity for eight elements commonly determined by flame AA. Matrix/Vol: 2% HCL/100 mL, Concentration of Element: 50mg/mL Calcium, Chromium, Copper, Iron, Nickel and 20mg/mL Potassium, and 10mg/mL Sodium and Zinc.
Catalog No. 50-211-3469 Supplier Perkin Elmer US LLC Supplier No. 02900540
Add to Cart
Add to Cart
Recommended for use in checking sensitivity for eight elements commonly determined by flame AA. Matrix/Vol: 2% HCL/100 mL, Concentration of Element: 50mg/mL Calcium, Chromium, Copper, Iron, Nickel and 20mg/mL Potassium, and 10mg/mL Sodium and Zinc.