Learn More
Medchemexpress LLC Aaa-10 | 2758171-70-7 | 98.0% | 472.65 g/mol | C25H41FO5S | 5 MG
SDP
Catalog No. 5000221445
Encompass_Preferred
AAA-10 is an orally active small-molecule inhibitor of gut bacterial bile salt hydrolases (BSH) used in research to probe microbial bile acid metabolism and downstream host effects.
  • Potent BSH inhibitor with IC50s of 10 nM (B. theta rBSH) and 80 nM (B. longum rBSH).
  • Inhibition in bacterial cultures: 74 nM (Gram-negative) and 901 nM (Gram-positive).
  • High purity (98.0%) and defined molecular weight (472.65 g/mol).
  • Soluble in DMSO (50 mg/mL) and compatible with several in vitro formulations.
  • Powder stability: -20°C up to 3 years, 4°C up to 2 years; in solvent stability varies.
  • Suitable for in vitro and in vivo formulation studies of bile acid metabolism.
  • Available in small solid quantities for research use.
Catalog No. 50-002-21445 Supplier Medchemexpress LLC Supplier No. HY1451475MG
AAA-10 is an orally active small-molecule inhibitor of gut bacterial bile salt hydrolases (BSH) used in research to probe microbial bile acid metabolism and downstream host effects.
  • Potent BSH inhibitor with IC50s of 10 nM (B. theta rBSH) and 80 nM (B. longum rBSH).
  • Inhibition in bacterial cultures: 74 nM (Gram-negative) and 901 nM (Gram-positive).
  • High purity (98.0%) and defined molecular weight (472.65 g/mol).
  • Soluble in DMSO (50 mg/mL) and compatible with several in vitro formulations.
  • Powder stability: -20°C up to 3 years, 4°C up to 2 years; in solvent stability varies.
  • Suitable for in vitro and in vivo formulation studies of bile acid metabolism.
  • Available in small solid quantities for research use.