Learn More
Medchemexpress LLC AAA-10 | 2758171-70-7 | 98.0% | C25H41FO5S | 50 MG
SDP
Catalog No. 5000353363
Encompass_Preferred
AAA-10 is an orally active gut bacterial bile salt hydrolases (BSH) inhibitor, with IC50s of 10 nM against B. theta rBSH and 80 nM against B. longum rBSH.
  • Inhibits BSH activity in bacterial cultures.
  • Significantly inhibits deconjugation of glycochenodeoxycholic acid-d4 or taurocholic acid-d4 substrates.
  • Decreases the abundances of deoxycholic acid (DCA) and lithocholic acid (LCA) in mice feces.
  • Displays high colonic exposure and low gut permeability.
Catalog No. 50-003-53363 Supplier Medchemexpress LLC Supplier No. HY14514750MG
AAA-10 is an orally active gut bacterial bile salt hydrolases (BSH) inhibitor, with IC50s of 10 nM against B. theta rBSH and 80 nM against B. longum rBSH.
  • Inhibits BSH activity in bacterial cultures.
  • Significantly inhibits deconjugation of glycochenodeoxycholic acid-d4 or taurocholic acid-d4 substrates.
  • Decreases the abundances of deoxycholic acid (DCA) and lithocholic acid (LCA) in mice feces.
  • Displays high colonic exposure and low gut permeability.