Learn More
Grainger RAYOVAC Battery, AAA
Catalog No. NC2132006
Encompass_Preferred
The Rayovac AAA alkaline battery is a single use, non-rechargeable battery with 1.5 volts. It has an estimated 10 year shelf life and is leak resistant. The battery measures 44.5 mm in height and 10.5 mm in diameter. It is sold individually with 6 batteries per display box. The battery is designed for low-drain devices like clocks, remote controls, toys and more. It is made in the USA.
This item is not returnable. View return policy
Catalog No. NC2132006 Supplier Grainger Supplier No. 64NU57
The Rayovac AAA alkaline battery is a single use, non-rechargeable battery with 1.5 volts. It has an estimated 10 year shelf life and is leak resistant. The battery measures 44.5 mm in height and 10.5 mm in diameter. It is sold individually with 6 batteries per display box. The battery is designed for low-drain devices like clocks, remote controls, toys and more. It is made in the USA.