Learn More
MSC Simpson AAA Triplex Pump Kit, 3200 PSI, 2.8 GPM, Brass Head
Catalog No. 502838137
Encompass
The AAA Triplex Pump Kit is an industrial pump designed for gas pressure washers. It delivers 3200 PSI at 2.8 GPM and is an easy-to-install replacement for models with a horizontal engine and 3/4 inch diameter shaft.
  • Easy installation for gas pressure washers with horizontal 3/4 inch shaft engines.
  • Dual oversized ball bearings for extended life.
  • Stainless steel unloader with easy-start system.
  • PowerBoost technology for high pressure, with adjustable power.
  • Sight glass and advanced oil seal for oil monitoring.
  • SAE standard bolt pattern ensures wide compatibility.
  • Includes siphon tube, mounting bolts, shaft key, and manual.
  • Durable brass head and black aluminum pump.
Catalog No. 50-283-8137 Supplier MSC Supplier No. 49039977
The AAA Triplex Pump Kit is an industrial pump designed for gas pressure washers. It delivers 3200 PSI at 2.8 GPM and is an easy-to-install replacement for models with a horizontal engine and 3/4 inch diameter shaft.
  • Easy installation for gas pressure washers with horizontal 3/4 inch shaft engines.
  • Dual oversized ball bearings for extended life.
  • Stainless steel unloader with easy-start system.
  • PowerBoost technology for high pressure, with adjustable power.
  • Sight glass and advanced oil seal for oil monitoring.
  • SAE standard bolt pattern ensures wide compatibility.
  • Includes siphon tube, mounting bolts, shaft key, and manual.
  • Durable brass head and black aluminum pump.