Learn More
ACROBiosystems Human IL-2RB&IL-2RA&IL-2RG, His Tag&Twin Strep Tag (ILG-H52W9) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (IL-2RB) & Glu 22 - Ser 212 (IL-2RA) & Leu 23 - Asn 254 (IL-2RG) (Accession # P14784-1 (IL-2RB) & P01
Catalog No. 502253143
Encompass_Preferred
Both Interleukin-2 receptor subunit beta and Interleukin-2 receptor subunit gamma are receptor for interleukin-2. Common subunit for the receptors for a variety of interleukins. Interacts with SHB upon interleukin stimulation. Probably in association with IL15RA, involved in the stimulation of neutrophil phagocytosis by IL15. This beta subunit is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2. IL2R exists in 3 different forms: a high affinity dimer, an intermediate affinity monomer (beta subunit), and a low affinity monomer (alpha subunit). The high and intermediate affinity forms also associate with a gamma subunit.
This item is not returnable. View return policy
Catalog No. 50-225-3143 Supplier ACROBiosystems Supplier No. ILGH52W9100UG
Both Interleukin-2 receptor subunit beta and Interleukin-2 receptor subunit gamma are receptor for interleukin-2. Common subunit for the receptors for a variety of interleukins. Interacts with SHB upon interleukin stimulation. Probably in association with IL15RA, involved in the stimulation of neutrophil phagocytosis by IL15. This beta subunit is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2. IL2R exists in 3 different forms: a high affinity dimer, an intermediate affinity monomer (beta subunit), and a low affinity monomer (alpha subunit). The high and intermediate affinity forms also associate with a gamma subunit.