Learn More
Sino Biological AACS / Acetoacetyl-CoA Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11117MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human AACS (rh AACS; Catalog#11117-H07B; NP_076417.2; Met 1-Phe 672).The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8044