Learn More
Aviva Systems Biology Corp Aacs Antibody - middle region (ARP60814_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60814_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Aacs. It was validated on Western Blot. . Alternate names: 2210408B16Rik, SUR5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-287-175