Learn More
Cayman Chemical aActInIn4NTRm RabbIt M 100uL
SDP
Catalog No. 502949216
Encompass_Preferred
An internal standard for the quantification of PGB2 by GC- or LC-MS.
Catalog No. 50-294-9216 Supplier Cayman Chemical Supplier No. 32239100 UL
An internal standard for the quantification of PGB2 by GC- or LC-MS.