Learn More
Aviva Systems Biology Corp AADAC Antibody - C-terminal region (ARP45038_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45038_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AADAC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AADAC. Alternate names: DAC, CES5A1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-58956