Learn More
Aviva Systems Biology Corp AADAC Antibody - N-terminal region (ARP45036_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45036_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AADAC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AADAC. Alternate names: CES5A1, DACApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58955