Learn More
Aviva Systems Biology Corp Aadacl1 Antibody - middle region (ARP49569_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49569_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Aadacl1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Aadacl1, B230106I24Rik, CPO-BP, NcehApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-436