Learn More
Aviva Systems Biology Corp AADACL3 Antibody - N-terminal region (ARP63127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AADACL3. It was validated on Western Blot. . Alternate names: RP11-474O21.3ApplicationWB. Species reactivity: Human
Catalog No. 50-287-888