Learn More
Aviva Systems Biology Corp AADACL4 Antibody - N-terminal region (ARP44654_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44654_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AADACL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AADACL4. Alternate names: -ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58864