Learn More
Enzo Life Sciences AAF-CMK (25mg). CAS: 184901-82-4
SDP
Supplier:  Enzo Life Sciences BMLPI1230025
Encompass_Preferred
A serine protease inhibitor which irreversibly inhibits tripeptidyl peptidase II (TPPII), a giant protease which may substitute for some proteasome functions, and reversibly inhibits TPPI. Shows only marginal inhibition of the chymotrypsin-like activity of the proteasome. Alternative name: Ala-Ala-Phe-CMK. Purity: ≥95% (TLC). Long Term Storage: -20°C.
Catalog No. 50-201-1461