Learn More
MSC AAF Flanders Pre-Pleat, 20x30x2 in Synthetic, MERV 8, 1 ea
Catalog No. 5000503968
Encompass
This pleated air filter is engineered for residential and light commercial HVAC systems, offering superior dust holding capacity and low pressure drop. It features 100% synthetic non-woven media and a robust, moisture-resistant cardboard frame, ensuring efficient air filtration and system protection. The filter is UL 900 classified.
  • Highest dust holding capacity and lowest pressure drop
  • UL 900 classified for safety
  • Synthetic non-woven, recyclable media
  • Expanded metal provides pleat stability and eliminates flutter
  • Heavy-duty, moisture-resistant frame with support members
  • Interlocking corners and positive media-to-frame seal prevent air-bypass
  • Filters smoke, pollen, bacteria, dust/lint, and mold spores
Catalog No. 50-005-03968 Supplier MSC Supplier No. 02739993
This pleated air filter is engineered for residential and light commercial HVAC systems, offering superior dust holding capacity and low pressure drop. It features 100% synthetic non-woven media and a robust, moisture-resistant cardboard frame, ensuring efficient air filtration and system protection. The filter is UL 900 classified.
  • Highest dust holding capacity and lowest pressure drop
  • UL 900 classified for safety
  • Synthetic non-woven, recyclable media
  • Expanded metal provides pleat stability and eliminates flutter
  • Heavy-duty, moisture-resistant frame with support members
  • Interlocking corners and positive media-to-frame seal prevent air-bypass
  • Filters smoke, pollen, bacteria, dust/lint, and mold spores