Learn More
Cayman Chemical AAFCMKtrIfluoroACTAte sal 10mg
SDP
Catalog No. 502971092
Encompass_Preferred
An irreversible inhibitor of TPPII commonly used at 10-100 µM; does not significantly interfere with the chymotrypsin-like activity of the proteasome; also inhibits bleomycin hydrolase and puromycin-sensitive aminopeptidase when used at 50 µM
Catalog No. 50-297-1092 Supplier Cayman Chemical Supplier No. 1446110 MG
An irreversible inhibitor of TPPII commonly used at 10-100 µM; does not significantly interfere with the chymotrypsin-like activity of the proteasome; also inhibits bleomycin hydrolase and puromycin-sensitive aminopeptidase when used at 50 µM