Learn More
Aviva Systems Biology Corp AAK1 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP55102_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AAK1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human AAK1. Alternate names: DKFZp686F03202, DKFZp686K16132, FLJ23712, FLJ25931, FLJ31060, FLJ42882, FLJ45252, KIAA1048, MGC138170, MGC164568, MGC164570ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-005