Learn More
Aviva Systems Biology Corp Aak1 Antibody - C-terminal region (ARP55101_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55101_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Aak1. It was validated on Western Blot. . Alternate names: 5530400K14Rik, 9630042K20, AU067724, AU067726, BC028270, C79663, D6Ertd245e, MGC40785, R75501, mKIAA1048ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-004