Learn More
Enzo Life Sciences AAL-993 (5mg). CAS: 269390-77-4
SDP
Supplier:  Enzo Life Sciences BMLEI3670005
Encompass_Preferred
Highly potent inhibitor of VEGFR-1 (IC50=130nM), VEGFR-2 (IC50=23nM) and VEGFR-3 (IC50=18nM). At higher concentrations it inhibits PDGFR-β (640nM), c-Kit (236nM) and CSF-1R (380nM). Alternative name: 2-[(4-Pyridyl)methyl]amino-N-[3-(trifluoromethyl)phenyl]benzamide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25 mg/ml) or ethanol (15 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0749