Learn More
Perkin Elmer US LLC Single Slot Burner Head for PerkinElmer AAnalyst Series Spectrometers, Slot Length: 10cm, Flame Type: Air-Acetylene
Catalog No. 509043598
Encompass_Preferred
The 10 cm burner head is designed to be used with the air-acetylene flame. Because of its long burner path length, it provides the best sensitivity for air-acetylene elements.
Catalog No. 50-904-3598 Supplier Perkin Elmer US LLC Supplier No. N0400102
The 10 cm burner head is designed to be used with the air-acetylene flame. Because of its long burner path length, it provides the best sensitivity for air-acetylene elements.