Learn More
Perkin Elmer US LLC Single Slot Burner Head for PerkinElmer AAnalyst Series Spectrometers, Slot Length: 5cm, Flame Type: Air-Acetylene
Catalog No. 509043597
Encompass_Preferred
A 5 cm air-acetylene burner head is available for applications in which reduced sensitivity is required. It can be rotated to provide further sensitivity reduction, and it has a wide slot to prevent clogging.
Catalog No. 50-904-3597 Supplier Perkin Elmer US LLC Supplier No. N0400101
A 5 cm air-acetylene burner head is available for applications in which reduced sensitivity is required. It can be rotated to provide further sensitivity reduction, and it has a wide slot to prevent clogging.